L-(-)-tert-Leucine (H-Tle-OH) 99%

Synonyms: (S)-2-Amino-3,3-dimethylbutanoic acidH-Tle-OH

AC29778-0010 AC29778-0050
200055-794EA 36.6 USD
200055-794 200006-862
L-(-)-tert-Leucine (H-Tle-OH) 99%
L(-)-tert-Leucine (H-Tle-OH)
Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064218
CAS Number: 20859-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR