(Chloromethylene)dimethylammonium chloride 96%

Danger

Synonyms: N-Chloromethylen-N,N-Dimethyl Ammonium Chloride

AC29577-0050 AC29577-0250
103532-146EA 42.9 USD
103532-146 200008-190
(Chloromethylene)dimethylammonium chloride 96%
(Chloromethylene)dimethylammonium chloride
Formula: C₃H₇Cl₂N
Melting Pt: 139 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011868
CAS Number: 3724-43-4
EINECS: 425-970-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR