2-Ethynylpyridine 98+%

AC29567-0010 AC29567-0250 AC29567-0050
103512-158EA 488 USD
103512-158 200022-650 103512-156
2-Ethynylpyridine 98+%
2-Ethynylpyridine
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 77…78 °C (14 mmHg)
Density: 1.02 g/cm³
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041598
CAS Number: 1945-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR