3-Phenyl-1H-pyrazole-4-carbaldehyde 98%

Warning

Synonyms: 3-Phenyl-1H-pyrazole-4-carboxaldehyde

AC29600-0010
200010-072EA 136 USD
200010-072
3-Phenyl-1H-pyrazole-4-carbaldehyde 98%
3-Phenyl-1H-pyrazole-4-carbaldehyde

Appearance: Light yellow Powder

Formula: C₁₀H₈N₂O
MW: 172.187 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00216971
CAS Number: 26033-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR