3-Methyl-1-phenyl-1H-pyrazole 98%

Warning

AC29539-0025
200047-516EA 37.2 USD
200047-516
3-Methyl-1-phenyl-1H-pyrazole 98%
3-Methyl-1-phenyl-1H-pyrazole
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Melting Pt: 33…37 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003123
CAS Number: 1128-54-7
EINECS: 214-438-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR