2,6-Diaminopurine 98%

Warning

Synonyms: 9H-Purine-2,6-diamine

AC29527-0050
200022-286EA 101 USD
200022-286
2,6-Diaminopurine 98%
2,6-Diaminopurine
Formula: C₅H₆N₆
MW: 150.14 g/mol
Melting Pt: 302 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00213668
CAS Number: 1904-98-9
EINECS: 217-605-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR