2,2-Dimethyl-4-pentenal 97% stabilized

Danger

AC29504-0050
200063-596EA 181 USD
200063-596
2,2-Dimethyl-4-pentenal 97% stabilized
2,2-Dimethyl-4-pentenal

Appearance: Clear colorless to light yellow Liquid

Formula: C₇H₁₂O
MW: 112.172 g/mol
Boiling Pt: 124…125 °C
Density: 0.825 g/cm³
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010699
CAS Number: 5497-67-6
UN: 1989
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR