7-Methoxy-2-tetralone 95%
Synonyms:
3,4-Dihydro-7-methoxynaphthalen-2(1H)-one
, 7-Methoxy-3,4-dihydronaphthalen-2(1H)-one
AC29497-0250
AC29497-0050
200024-208EA
149.7
USD
200024-208
103512-144
7-Methoxy-2-tetralone 95%
7-Methoxy-2-tetralone
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Boiling Pt: 124…126 °C (1.5 mmHg) Density: 1.13 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001730 CAS Number: 4133-34-0 EINECS: 223-954-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...