(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 99%

Danger

Synonyms: (1S,2S)-N,N'-Dimethyl-1,2-diphenylethane-1,2-diamine

AC29467-2500
200033-274EA 288.6 USD
200033-274
(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 99%
(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine

Appearance: White to light yellow Crystalline powder or crystals

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Melting Pt: 48…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00216956
CAS Number: 70749-06-3
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR