(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 97%

Warning

Synonyms: Methyl N-(Triethylammoniosulphonyl)carbamateBurgess ReagentMethyl N-(Triethylammoniosulfonyl)carbamate

AC29451-0250 AC29451-0010 AC29451-0050
200063-590EA 267 USD
200063-590 103517-784 200003-054
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 97%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt
Formula: C₈H₁₈N₂O₄S
MW: 238.30 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Freezer
MDL Number: MFCD00077815
CAS Number: 29684-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR