1,4,7-Triazacyclononane trihydrochloride 97%

Danger

Synonyms: 1,4,7-Triazonane trihydrochloride

AC29431-0025 AC29431-5000
103532-120EA 462 USD
103532-120 200021-664
1,4,7-Triazacyclononane trihydrochloride 97%
1,4,7-Triazacyclononane trihydrochloride
Formula: C₆H₁₈Cl₃N₃
MW: 238.5900 g/mol
Melting Pt: 285…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00074887
CAS Number: 58966-93-1
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR