6-Methylbenzo[d]oxazole 99.5+%

Warning

Synonyms: 6-Methylbenzoxazole

AC29311-0100 AC29311-0025
103512-104EA 355.4 USD
103512-104 200010-818
6-Methylbenzo[d]oxazole 99.5+%
6-Methylbenzo[d]oxazole
Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00216937
CAS Number: 10531-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR