Dimcarb (dimethylammonium dimethylcarbamate) (Carbon dioxide:Dimethylamine) <2:1

Danger

Synonyms: N-Methylmethanamine dimethylcarbamateDimethylamine carbon dioxide complex

AC29290-1000 AC29290-0250
103527-344EA 31.3 USD
103527-344 200031-036
Dimcarb (dimethylammonium dimethylcarbamate) (Carbon dioxide:Dimethylamine) <2:1
Dimcarb (Dimethylammonium dimethylcarbamate)
Formula: (CH₃)₂NH · (CH₃)₂NCOOH
MW: 134.18 g/mol
Boiling Pt: 60…61 °C (760 mmHg)
Density: 1.05 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00029888
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR