5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 95%

Warning

Synonyms: 5,10,15,20-Tetrakis(perfluorophenyl)porphyrin5,10,15,20-Tetrakis(pentafluorophenyl)-21h,23h-porphine5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine

AC29215-1000
200024-012EA 103 USD
200024-012
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 95%
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

Appearance: Violet to purple Crystalline powder

Formula: C₄₄H₁₀F₂₀N₄
MW: 974.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010032
CAS Number: 25440-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR