6-Carbethoxy-2,2,6-trimethylcyclohexanone ≥97%

AC29189-0010
200036-326EA 83.2 USD
200036-326
6-Carbethoxy-2,2,6-trimethylcyclohexanone ≥97%
6-Carbethoxy-2,2,6-trimethylcyclohexanone

Appearance: Clear colorless Liquid

Formula: C₁₂H₂₀O₃
Boiling Pt: 55…56 °C (3 mmHg)
Storage Temperature: Ambient
CAS Number: 7507-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR