1-Cyclohexene-1-carboxylic acid 96%

Warning

Synonyms: Cyclohex-1-enecarboxylic acid

AC29175-0025
200021-808EA 93.7 USD
200021-808
1-Cyclohexene-1-carboxylic acid 96%
1-Cyclohexene-1-carboxylic acid
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 137 °C (15 mmHg)
Melting Pt: 35…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00001545
CAS Number: 636-82-8
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR