2-Ethylhexyl-4-methoxycinnamate 98% stabilized

Synonyms: Octinoxate4-Methoxycinnamic Acid Octyl EsterOctyl 4-Methoxycinnamate4-Methoxycinnamic Acid 2-Ethylhexyl Ester2-Ethylhexyl 3-(4-methoxyphenyl)acrylate

AC29116-1000 AC29116-0250
103512-076EA 163 USD
103512-076 200003-022
2-Ethylhexyl-4-methoxycinnamate 98% stabilized
2-Ethylhexyl-4-methoxycinnamate
Formula: C₁₈H₂₆O₃
MW: 290.40 g/mol
Density: 1.009 g/cm³
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00072582
CAS Number: 5466-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR