(+)-2-Methyl-L-serine 99%

Synonyms: (S)-2-Amino-3-hydroxy-2-methylpropanoic acid

AC29001-2500
200021-376EA 329 USD
200021-376
(+)-2-Methyl-L-serine 99%
(+)-2-Methyl-L-serine

Appearance: White Powder

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 286…288 °C
Storage Temperature: Ambient
MDL Number: MFCD00145243
CAS Number: 16820-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR