2-Chloro-2-ethoxyacetic acid ethyl ester 90% remainder mainly ethyl diethoxyacetate

Warning

Synonyms: Ethyl chloro(ethoxy)acetate

AC27987-0500
200056-868EA 246 USD
200056-868
2-Chloro-2-ethoxyacetic acid ethyl ester 90% remainder mainly ethyl diethoxyacetate
2-Chloro-2-ethoxyacetic acid ethyl ester

Appearance: Clear colorless to orange-brown Liquid

Formula: C₆H₁₁ClO₃
MW: 166.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043953
CAS Number: 34006-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR