3-Nitro-1,8-naphthalic anhydride 98%

Warning

Synonyms: 5-Nitrobenzo[de]isochromene-1,3-dione

AC27873-0250 AC27873-0050
200041-658EA 45.8 USD
200041-658 200017-174
3-Nitro-1,8-naphthalic anhydride 98%
3-Nitro-1,8-naphthalic anhydride
Formula: C₁₂H₅NO₅
MW: 243.1718 g/mol
Melting Pt: 247…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00006926
CAS Number: 3027-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR