Brucine, anhydrous 99%

Danger

Synonyms: 2,3-Dimethoxystrychnidin-10-one10,11-Dimethoxystrychnine

AC27872-0050
103527-286EA 25.1 USD
103527-286
Brucine, anhydrous 99%
Brucine
Formula: C₂₃H₂₆N₂O₄
MW: 394.47 g/mol
Melting Pt: 174…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00005942
CAS Number: 357-57-3
UN: 1570
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR