Geranyl linalool mixture of isomers ∼95% tech.

Warning

AC27836-0250 AC27836-0025
200037-518EA 166 USD
200037-518 103509-830
Geranyl linalool mixture of isomers ∼95% tech.
Geranyl linalool
Formula: C₂₀H₃₄O
Boiling Pt: 250 °C (760 mmHg)
Density: 0.88 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00059363
CAS Number: 1113-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR