4'-Carboxybenzo-18-crown-6 (4'-carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane) 99%
|
Warning
|
Synonyms:
2,3,5,6,8,9,11,12,14,15-Decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine-18-carboxylic acid
AC27749-0010
200023-674EA
201.4
USD
200023-674
4'-Carboxybenzo-18-crown-6 (4'-carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane) 99%
4'-Carboxybenzo-18-crown-6 (4'-Carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...