1-Methyl-2-benzimidazolinone 98%

Warning

Synonyms: 1-Methyl-1H-benzo[d]imidazol-2(3H)-one1,3-Dihydro-1-methyl-2H-benzimidazol-2-one

AC27650-0050
200008-708EA 177 USD
200008-708
1-Methyl-2-benzimidazolinone 98%
1-Methyl-2-benzimidazolinone

Appearance: White to light beige Crystalline powder

Formula: C₈H₈N₂O
MW: 148.165 g/mol
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00142654
CAS Number: 1849-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR