1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%

Danger

Synonyms: Diaza-18-crown-61,4,10,13-Tetraoxa-7,16-diazacyclooctadecane

AC27582-1000
103532-072EA 35 USD
103532-072
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%
1,10-Diaza-18-crown-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane, Kryptofix® 22)
Formula: C₁₂H₂₆N₂O₄
MW: 262.35 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005112
CAS Number: 23978-55-4
EINECS: 245-965-0
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR