N-Boc-L-valine 99+%

Synonyms: BOC-L-ValineBoc-Val-OH

AC27571-0250 AC27571-0050
200018-344EA 88.4 USD
200018-344 200030-600
N-Boc-L-valine 99+%
N-Boc-L-valine
Formula: C₁₀H₁₉NO₄
MW: 217.3 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065605
CAS Number: 13734-41-3
EINECS: 237-307-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR