2-Methyl-3-furanthiol ca.90% balance oxidized compound

Danger

Synonyms: 3-Mercapto-2-methylfuran

AC27539-0010 AC27539-0050
200063-550EA 204.9 USD
200063-550 200028-862
2-Methyl-3-furanthiol ca.90% balance oxidized compound
2-Methyl-3-furanthiol
Formula: C₅H₆OS
MW: 114.16 g/mol
Boiling Pt: 57…60 °C (44 mmHg)
Flash Pt: 37 °C
Storage Temperature: Freezer
MDL Number: MFCD00010280
CAS Number: 28588-74-1
UN: 3071
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR