ɑ-Hydroxy-γ-butyrolactone 99%

Warning

Synonyms: 3-Hydroxytetrahydrofuran-2-oneDihydro-3-hydroxy-2(3H)-furanone3-Hydroxydihydrofuran-2(3H)-one

AC27507-0050 AC27507-0250
200009-426EA 354 USD
200009-426 200022-718
ɑ-Hydroxy-γ-butyrolactone 99%
alpha-Hydroxy-gamma-butyrolactone

Appearance: Clear colorless to yellow Viscous liquid

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 85…90 °C (.8mbar)
Density: 1.313 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00134268
CAS Number: 19444-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR