3-Amino-5-hydroxy-1H-pyrazole 98%

Warning

Synonyms: 3-Amino-1H-pyrazol-5(4H)-one

AC27172-0250 AC27172-0050
200041-442EA 98.9 USD
200041-442 200029-182
3-Amino-5-hydroxy-1H-pyrazole 98%
3-Amino-5-hydroxy-1H-pyrazole
Formula: C₃H₅N₃O
MW: 99.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022384
CAS Number: 6126-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR