Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phenylglycine ≥93%

Supplier: ACROS ORGANICS MS


AC27608-1000 AC27608-0250
200050-282EA 123.84 USD InStock
200050-282 200026-246
N-Phenylglycine ≥93%
N-Phenylglycine


Order Now
Specification Test Results
Appearance Ochre to yellow-brown Powder
Infrared spectrum Conforms
Melting point 121°C to 127°C
HPLC ≥91 %
Titration with TBAH ≥91 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty