Zinc-copper couple

Warning

AC27144-1000 AC27144-0250
200015-170EA 256.31 USD
200015-170 200008-076
Zinc-copper couple
Zinc-copper couple

Appearance: Grey-black Powder

Formula: CuZn
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00084851
CAS Number: 53801-63-1
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR