N-Benzylglycine ethyl ester hydrochloride 98%

AC27137-0250
200026-100EA 55.7 USD
200026-100
N-Benzylglycine ethyl ester hydrochloride 98%
N-Benzylglycine ethyl ester hydrochloride

Appearance: White to beige Powder

Formula: C₁₁H₁₆ClNO₂
Melting Pt: 96…100 °C
Storage Temperature: Refrigerator
CAS Number: 6344-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR