α-Ketoglutaric acid disodium salt dihydrate 99%

Synonyms: α-Ketoglutaric acid disodium salt dihydrate2-Oxoglutaric acid disodium salt dihydrateSodium α-ketoglutarate dibasic dihydrate

AC27125-0500 AC27125-0050 AC27125-0010
103518-700EA 847.5 USD
103518-700 200041-102 200041-104
α-Ketoglutaric acid disodium salt dihydrate 99%
α-Ketoglutaric acid disodium salt dihydrate
Formula: C₅H₄O₅·2Na·2H₂O
MW: 226.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150702
CAS Number: 305-72-6
EINECS: 206-167-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR