1-Benzyl-3-pyrrolidinone 98%

Warning

Synonyms: 1-(Phenylmethyl)-3-pyrrolidinone1-Benzylpyrrolidin-3-one

AC27103-0025 AC27103-0100 AC27103-0010
200002-968EA 55.7 USD
200002-968 103511-984 103511-982
1-Benzyl-3-pyrrolidinone 98%
1-Benzyl-3-pyrrolidinone
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 109 °C (.1 mmHg)
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005342
CAS Number: 775-16-6
EINECS: 212-274-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR