1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%

Synonyms: 1,2:5,6-Di-o-isopropylidene-alpha-D-allofuranose

AC27096-0025 AC27096-5000 AC27096-0100
103511-978EA 318 USD
103511-978 200015-514 103511-980
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
Formula: C₁₂H₂₀O₆
MW: 260.28 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135634
CAS Number: 2595-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR