D(+)-α-Acetobromoglucose 98.5%
|
Warning
|
Synonyms:
Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose
, 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide
, Acetobromo glucose
, Acetobromo-α-D-glucose
AC27093-0100
AC27093-0500
103511-974EA
210
USD
103511-974
200004-854
D(+)-α-Acetobromoglucose 98.5%
D(+)-α-Acetobromoglucose
Formula:
C₁₄H₁₉BrO₉ MW: 411.2 g/mol Melting Pt: 83…88 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063254 EINECS: 209-339-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...