Methyl-β-D-glucopyranoside hemihydrate 99%

Warning

Synonyms: Methyl beta-D-Glucoside

AC27092-0250
200025-906EA 241 USD
200025-906
Methyl-β-D-glucopyranoside hemihydrate 99%
Methyl β-D-glucopyranoside hemihydrate
Formula: C₇H₁₄O₆·0.5H₂O
MW: 203.19 g/mol
Melting Pt: 111…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149442
EINECS: 211-909-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR