2-Chloro-6-methylnicotinic acid 98+%

AC27081-0250 AC27081-0050
200016-476EA 190 USD
200016-476 200028-638
2-Chloro-6-methylnicotinic acid 98+%
2-Chloro-6-methylnicotinic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS Number: 30529-70-5
EINECS: 250-229-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR