Dimethyl methoxymethylenemalonate 98+%

Warning

Synonyms: Methoxymethylenemalonic Acid Dimethyl EsterDimethyl Methoxymethylenemalonate

AC26956-0250 AC26956-2500
200025-062EA 258 USD
200025-062 200012-104
Dimethyl methoxymethylenemalonate 98+%
Dimethyl methoxymethylenemalonate

Appearance: White Crystalline solid

Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 98 °C (.5 mmHg)
Melting Pt: 40…44 °C
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00038225
CAS Number: 22398-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR