Potassium methyl malonate 98%

Warning

Synonyms: Monomethyl monopotassium malonateMonomethyl malonate potassium salt

AC26955-0250 AC26955-5000 AC26955-1000 AC26955-0050
200007-052EA 55.8 USD
200007-052 103511-940 200019-840 103511-938
Potassium methyl malonate 98%
Potassium methyl malonate
Formula: C₄H₅KO₄
MW: 156.18 g/mol
Melting Pt: >200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014021
CAS Number: 38330-80-2
EINECS: 253-886-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR