Ethyl-3-ethoxypropionate 99+% stabilized, pure

Warning

Synonyms: Ethyl 3-ethoxypropanoate

AC26949-0025 AC26949-0010 AC26949-0100
103532-042EA 336.92 USD
103532-042 200012-360 200006-454
Ethyl-3-ethoxypropionate 99+% stabilized, pure
Ethyl 3-ethoxypropionate
Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 170 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.949 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
EINECS: 212-112-9
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR