Acetylsalicylsalicylic acid 97%

Warning

Synonyms: 2-((2-Acetoxybenzoyl)oxy)benzoic acid

AC26935-2500 AC26935-0010
200024-300EA 205 USD
200024-300 103509-956
Acetylsalicylsalicylic acid 97%
Acetylsalicylsalicylic acid
Formula: C₁₆H₁₂O₆
MW: 300.266 g/mol
Melting Pt: 145…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00143537
CAS Number: 530-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR