Triphenylphosphonium bromide 99%
|
Danger
|
Synonyms:
Triphenylphosphine hydrobromide
AC26909-0250
AC26909-1000
AC26909-0050
200033-108EA
211
USD
200033-108
200021-090
103532-038
Triphenylphosphonium bromide 99%
Triphenylphosphonium bromide
Formula:
C₁₈H₁₆BrP Melting Pt: 196…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035107 CAS Number: 6399-81-1 EINECS: 229-012-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...