5-Methylfurfurylamine 98%

Danger

Synonyms: 2-Aminomethyl-5-methylfuran(5-Methylfuran-2-yl)methanamine5-Methyl-2-furanmethanamine

AC26894-0050 AC26894-0250
200024-112EA 166 USD
200024-112 200063-528
5-Methylfurfurylamine 98%
5-Methylfurfurylamine

Appearance: Clear yellow Liquid

Formula: C₆H₉NO
MW: 111.14 g/mol
Boiling Pt: 57 °C (13mbar)
Melting Pt: –22 °C
Density: 1.01 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00143471
CAS Number: 14003-16-8
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR