Tetramethylsilane 99%

Danger

Synonyms: TMS

AC26886-5000 AC26886-1000
102777-060EA 285 USD
102777-060 102775-172
Tetramethylsilane 99%
Tetramethylsilane
Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26.6 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.64 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR