Rhodium(II) acetate dimer (∼46% Rh), anhydrous

Warning

Synonyms: Tetrakis(acetato)dirhodium(II)Dirhodium tetraacetate

AC26863-0050
103518-680EA 3099 USD
103518-680
Rhodium(II) acetate dimer (∼46% Rh), anhydrous
Rhodium (II) acetate dimer
Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00003538
CAS Number: 15956-28-2
EINECS: 240-084-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR