(-)-Terpinen-4-ol 97% (sum of enantiomers)

Warning

Synonyms: (-)-4-Hydroxy-4-isopropyl-1-methyl-1-cyclohexene(-)-p-Menth-1-en-4-ol(-)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol(R)-1-Isopropyl-4-methylcyclohex-3-enol

AC27043-1000
200033-214EA 245.77 USD
200033-214
(-)-Terpinen-4-ol 97% (sum of enantiomers)
(-)-Terpinen-4-ol

Appearance: Clear colorless to yellow Liquid

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 212 °C (760 mmHg)
Density: 0.933 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00167108
CAS Number: 20126-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR