Chlortetracycline hydrochloride can be used as secondary standard

Warning

Synonyms: 7-Chlorotetracycline hydrochlorideAureomycin

AC26824-5000 AC26824-0025
103509-940EA 433.66 USD
103509-940 200006-026
Chlortetracycline hydrochloride can be used as secondary standard
Chlortetracycline hydrochloride
Formula: C₂₂H₂₃ClN₂O₈·HCl
MW: 515.34 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD26142699
CAS Number: 64-72-2
EINECS: 200-591-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR