2-Amino-1,3,4-thiadiazole 97%

Danger

Synonyms: 1,3,4-Thiadiazol-2-amine[1,3,4]Thiadiazol-2-ylamine

AC26816-0010
103532-034EA 24.8 USD
103532-034
2-Amino-1,3,4-thiadiazole 97%
2-Amino-1,3,4-thiadiazole
Formula: C₂H₃N₃S
MW: 101.13 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00003107
CAS Number: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR