2-(Methyldithio)isobutyraldehyde 95%

Warning

AC26515-0010 AC26515-0050
103509-928EA 648 USD
103509-928 200002-904
2-(Methyldithio)isobutyraldehyde 95%
2-(Methyldithio)isobutyraldehyde
Formula: C₅H₁₀OS₂
Boiling Pt: 46 °C (1 mmHg)
Density: 1.1 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00142943
CAS Number: 67952-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR